siRNA Details
| Gene Symbol: Rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005889.2 | NP_001005889.2 | 8 | 1031 | ||
| GenBank mRNA | AY428868.1 | - | 7 | 856 | ||
| BC087147.1 | - | 8 | 1031 | 7130553 | ||
| FQ218538.1 | - | 5 | 700 | |||
| KF307742.1 | - | 7 | 874 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009041, NM_001104617, NM_001104616, AK206105, X60672, and others.

Rdx
Genomic Map