siRNA Details
| Gene Symbol: Zc3h12c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108146.1 | NP_001101616.1 | 2 | 343 | ||
| XM_006243073.2 | XP_006243135.1 | 2 | 563 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h12c| Gene Symbol: Zc3h12c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108146.1 | NP_001101616.1 | 2 | 343 | ||
| XM_006243073.2 | XP_006243135.1 | 2 | 563 | |||