siRNA Details
| Gene Symbol: Exph5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_236272.6 | XP_236272.4 | 2, 3 | 525 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exph5| Gene Symbol: Exph5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_236272.6 | XP_236272.4 | 2, 3 | 525 | ||