siRNA Details
| Gene Symbol: Dmxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766357.1 | XP_008764579.1 | 16 | 3185 | ||
| XM_008766358.1 | XP_008764580.1 | 16 | 3185 | |||
| XM_008766359.1 | XP_008764581.1 | 16 | 3185 | |||
| XM_008766360.1 | XP_008764582.1 | 16 | 3185 | |||

Dmxl2
Genomic Map