siRNA Details
| Gene Symbol: Hmg20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108150.1 | NP_001101620.1 | 2, 3 | 361 | ||
| XM_006243117.2 | XP_006243179.1 | 2, 3 | 3918 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmg20a| Gene Symbol: Hmg20a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108150.1 | NP_001101620.1 | 2, 3 | 361 | ||
| XM_006243117.2 | XP_006243179.1 | 2, 3 | 3918 | |||