siRNA Details
Gene Symbol: Hmg20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108150.1 | NP_001101620.1 | 4 | 561 | ||
XM_006243117.2 | XP_006243179.1 | 4 | 4118 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hmg20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108150.1 | NP_001101620.1 | 4 | 561 | ||
XM_006243117.2 | XP_006243179.1 | 4 | 4118 |