siRNA Details
| Gene Symbol: Snx33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127488.1 | NP_001120960.1 | 1 | 1454 | ||
| XM_006243123.2 | XP_006243185.1 | 2 | 2283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx33| Gene Symbol: Snx33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127488.1 | NP_001120960.1 | 1 | 1454 | ||
| XM_006243123.2 | XP_006243185.1 | 2 | 2283 | |||