siRNA Details
| Gene Symbol: RGD1305464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025011.2 | NP_001020182.2 | 3 | 3899 | ||
| GenBank mRNA | BC089935.1 | - | 2 | 973 | ||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 RGD1305464
RGD1305464| Gene Symbol: RGD1305464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025011.2 | NP_001020182.2 | 3 | 3899 | ||
| GenBank mRNA | BC089935.1 | - | 2 | 973 | ||
 Back To Top
 Back To Top