siRNA Details
| Gene Symbol: Rpp25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012124.1 | NP_001012124.1 | 1 | 571 | ||
| GenBank mRNA | BC087592.1 | - | 1 | 571 | 7309941 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpp25| Gene Symbol: Rpp25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012124.1 | NP_001012124.1 | 1 | 571 | ||
| GenBank mRNA | BC087592.1 | - | 1 | 571 | 7309941 | |