siRNA Details
| Gene Symbol: Csk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030039.1 | NP_001025210.1 | 5 | 549 | ||
| XM_006243163.1 | XP_006243225.1 | 6 | 1195 | |||
| XM_006243164.2 | XP_006243226.1 | 6 | 878 | |||
| GenBank mRNA | BC098863.1 | - | 5 | 549 | 7380519 | |
| X58631.1 | - | 6 | 584 | |||

Csk
Genomic Map