siRNA Details
| Gene Symbol: Kif23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108155.1 | NP_001101625.1 | 5, 6 | 553 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif23| Gene Symbol: Kif23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108155.1 | NP_001101625.1 | 5, 6 | 553 | ||