siRNA Details
| Gene Symbol: Paqr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014092.1 | NP_001014114.1 | 4 | 528 | ||
| XM_006243208.2 | XP_006243270.1 | 4 | 460 | |||
| XM_006243209.2 | XP_006243271.1 | 3 | 797 | |||
| XM_008766275.1 | XP_008764497.1 | 4 | 388 | |||
| XM_008766276.1 | XP_008764498.1 | 3 | 332 | |||
| GenBank mRNA | BC087040.1 | - | 4 | 528 | 7106223 | |
More Information
Set Membership: |
|

Paqr5
Genomic Map