siRNA Details
| Gene Symbol: Itga11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766278.1 | XP_008764500.1 | 4 | 725 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itga11| Gene Symbol: Itga11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766278.1 | XP_008764500.1 | 4 | 725 | ||