siRNA Details
| Gene Symbol: Plekho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_236354.5 | XP_236354.1 | 6 | 1578 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekho2| Gene Symbol: Plekho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_236354.5 | XP_236354.1 | 6 | 1578 | ||