siRNA Details
| Gene Symbol: Zfp609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173371.1 | NP_001166842.1 | 4 | 3024 | ||
| XM_008766321.1 | XP_008764543.1 | 4 | 5913 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp609| Gene Symbol: Zfp609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173371.1 | NP_001166842.1 | 4 | 3024 | ||
| XM_008766321.1 | XP_008764543.1 | 4 | 5913 | |||