siRNA Details
Gene Symbol: Zfp609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173371.1 | NP_001166842.1 | 4 | 2507 | ||
XM_008766321.1 | XP_008764543.1 | 4 | 5396 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp609 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173371.1 | NP_001166842.1 | 4 | 2507 | ||
XM_008766321.1 | XP_008764543.1 | 4 | 5396 |