siRNA Details
| Gene Symbol: Tln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766402.1 | XP_008764624.1 | 7, 8 | 893 | ||
| XM_008766403.1 | XP_008764625.1 | 7, 8 | 2931 | |||
| XM_008766404.1 | XP_008764626.1 | 7, 8 | 893 | |||
| XM_008766405.1 | XP_008764627.1 | 7, 8 | 893 | |||

Tln2
Genomic Map