siRNA Details
| Gene Symbol: Prtg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037651.1 | NP_001032740.1 | 7, 8 | 1182 | ||
| GenBank mRNA | AY630256.1 | - | 7, 8 | 1182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prtg| Gene Symbol: Prtg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037651.1 | NP_001032740.1 | 7, 8 | 1182 | ||
| GenBank mRNA | AY630256.1 | - | 7, 8 | 1182 | ||