siRNA Details
| Gene Symbol: Fam83b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108169.1 | NP_001101639.1 | 2 | 510 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam83b| Gene Symbol: Fam83b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108169.1 | NP_001101639.1 | 2 | 510 | ||