siRNA Details
| Gene Symbol: Elovl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191796.1 | NP_001178725.1 | 6 | 709 | ||
| XM_008766293.1 | XP_008764515.1 | 6 | 926 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elovl4| Gene Symbol: Elovl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191796.1 | NP_001178725.1 | 6 | 709 | ||
| XM_008766293.1 | XP_008764515.1 | 6 | 926 | |||