siRNA Details
| Gene Symbol: Ube3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039610.1 | NP_001034699.1 | 3 | 438 | ||
| XM_006243479.2 | XP_006243541.1 | 3 | 456 | |||
| GenBank mRNA | BC101916.1 | - | 3 | 438 | 7935691 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube3d| Gene Symbol: Ube3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039610.1 | NP_001034699.1 | 3 | 438 | ||
| XM_006243479.2 | XP_006243541.1 | 3 | 456 | |||
| GenBank mRNA | BC101916.1 | - | 3 | 438 | 7935691 | |