siRNA Details
| Gene Symbol: Prss35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008560.1 | NP_001008560.1 | 2 | 692 | ||
| XM_008766446.1 | XP_008764668.1 | 2 | 833 | |||
| GenBank mRNA | AB180912.1 | - | 2 | 692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss35| Gene Symbol: Prss35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008560.1 | NP_001008560.1 | 2 | 692 | ||
| XM_008766446.1 | XP_008764668.1 | 2 | 833 | |||
| GenBank mRNA | AB180912.1 | - | 2 | 692 | ||