siRNA Details
| Gene Symbol: Snx14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108174.1 | NP_001101644.1 | 6 | 661 | ||
| GenBank mRNA | BC160909.1 | - | 6 | 619 | 7455732 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snx14| Gene Symbol: Snx14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108174.1 | NP_001101644.1 | 6 | 661 | ||
| GenBank mRNA | BC160909.1 | - | 6 | 619 | 7455732 | |