siRNA Details
Gene Symbol: Tbc1d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108175.1 | NP_001101645.1 | 7 | 966 | ||
XM_006243528.2 | XP_006243590.1 | 6 | 1145 | |||
XM_008766449.1 | XP_008764671.1 | 6 | 1168 |
