siRNA Details
| Gene Symbol: RGD1309079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134472.1 | NP_001127944.1 | 2 | 1150 | ||
| GenBank mRNA | AY325189.1 | - | 273 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1309079| Gene Symbol: RGD1309079 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134472.1 | NP_001127944.1 | 2 | 1150 | ||
| GenBank mRNA | AY325189.1 | - | 273 | |||