siRNA Details
| Gene Symbol: U2surp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002729954.4 | XP_002730000.1 | 18 | 1855 | ||
| XM_006243554.1 | XP_006243616.1 | 19 | 2000 | |||
| XM_236501.7 | XP_236501.3 | 19 | 2041 | |||
| GenBank mRNA | FQ226547.1 | - | 19 | 2009 | ||

U2surp
Genomic Map