siRNA Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173349.1 | NP_001166820.1 | 2 | 812 | ||
| XM_008773419.1 | XP_008771641.1 | 2 | 885 | |||
| GenBank mRNA | BC160840.1 | - | 2 | 673 | 7460350 | |
| BC167061.1 | - | 2 | 812 | 8364382 | ||
| FQ210020.1 | - | 2 | 800 | |||
| FQ218340.1 | - | 2 | 800 | |||

Rnf128
Genomic Map