siRNA Details
| Gene Symbol: Vsig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037784.1 | NP_001032873.1 | 3 | 408 | ||
| XM_006234243.2 | XP_006234305.1 | 4 | 2731 | |||
| XM_006234244.2 | XP_006234306.1 | 3 | 2622 | |||
| XM_006234245.2 | XP_006234307.1 | 2 | 3059 | |||
| XM_006234246.2 | XP_006234308.1 | 2 | 1310 | |||
| GenBank mRNA | BC098943.1 | - | 3 | 408 | 7456980 | |

Vsig1
Genomic Map