siRNA Details
| Gene Symbol: Nme9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766584.1 | XP_008764806.1 | 5 | 326 | ||
| XM_008766585.1 | XP_008764807.1 | 5 | 325 | |||
| XM_008766586.1 | XP_008764808.1 | 5 | 326 | |||
| XM_008766587.1 | XP_008764809.1 | 5 | 325 | |||

Nme9
Genomic Map