siRNA Details
| Gene Symbol: Armc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173354.2 | NP_001166825.1 | 11, 12 | 1135 | ||
| XM_006243622.2 | XP_006243684.1 | 11, 12 | 1372 | |||
| XM_006243623.2 | XP_006243685.1 | 11, 12 | 1372 | |||
| XM_006243625.2 | XP_006243687.1 | 10, 11 | 1211 | |||
| GenBank mRNA | BC168192.1 | - | 11, 12 | 1043 | ||
| FQ211639.1 | - | 10, 11 | 1198 | |||

Armc8
Genomic Map