siRNA Details
| Gene Symbol: Cldn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014096.1 | NP_001014118.1 | 4, 5 | 643 | ||
| XM_006243629.2 | XP_006243691.1 | 4, 5 | 713 | |||
| GenBank mRNA | BC088427.1 | - | 4, 5 | 643 | 7124060 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn18| Gene Symbol: Cldn18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014096.1 | NP_001014118.1 | 4, 5 | 643 | ||
| XM_006243629.2 | XP_006243691.1 | 4, 5 | 713 | |||
| GenBank mRNA | BC088427.1 | - | 4, 5 | 643 | 7124060 | |