siRNA Details
| Gene Symbol: Stag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108179.1 | NP_001101649.1 | 7 | 858 | ||
| XM_006243631.2 | XP_006243693.1 | 7 | 853 | |||
| XM_006243632.2 | XP_006243694.1 | 8 | 870 | |||
| XM_008766528.1 | XP_008764750.1 | 8 | 829 | |||

Stag1
Genomic Map