siRNA Details
Gene Symbol: Stag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108179.1 | NP_001101649.1 | 26 | 3020 | ||
XM_006243631.2 | XP_006243693.1 | 29 | 3423 | |||
XM_006243632.2 | XP_006243694.1 | 30 | 3440 | |||
XM_008766528.1 | XP_008764750.1 | 30 | 3399 |
