siRNA Details
| Gene Symbol: Nek11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108182.1 | NP_001101652.1 | 4, 5 | 966 | ||
| XM_008766530.1 | XP_008764752.1 | 3, 4 | 600 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nek11| Gene Symbol: Nek11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108182.1 | NP_001101652.1 | 4, 5 | 966 | ||
| XM_008766530.1 | XP_008764752.1 | 3, 4 | 600 | |||