siRNA Details
| Gene Symbol: Rbm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108186.1 | NP_001101656.1 | 3 | 636 | ||
| XM_006243780.2 | XP_006243842.1 | 4 | 732 | |||
| XM_006243781.2 | XP_006243843.1 | 3 | 905 | |||
| XM_008766534.1 | XP_008764756.1 | 3 | 871 | |||

Rbm6
Genomic Map