siRNA Details
Gene Symbol: Rbm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108186.1 | NP_001101656.1 | 9, 10 | 2209 | ||
XM_006243780.2 | XP_006243842.1 | 10, 11 | 2305 | |||
XM_006243781.2 | XP_006243843.1 | 9, 10 | 2478 | |||
XM_008766534.1 | XP_008764756.1 | 9, 10 | 2444 | |||
XM_008766535.1 | XP_008764757.1 | 4, 5 | 1174 |
