siRNA Details
| Gene Symbol: Arih2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012275.2 | NP_001012275.1 | 12 | 1183 | ||
| XM_006243784.2 | XP_006243846.1 | 12 | 1651 | |||
| XM_006243785.2 | XP_006243847.1 | 12 | 1648 | |||
| XM_006243787.2 | XP_006243849.1 | 11 | 872 | |||
| XM_008766537.1 | XP_008764759.1 | 12 | 1336 | |||
| XM_008766538.1 | XP_008764760.1 | 12 | 1339 | |||
| GenBank mRNA | CV126331.1 | - | 8 | 687 | ||

Arih2
Genomic Map