siRNA Details
| Gene Symbol: Ccdc51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014098.1 | NP_001014120.1 | 4 | 1007 | ||
| XM_006243788.2 | XP_006243850.1 | 4 | 1211 | |||
| XM_006243789.2 | XP_006243851.1 | 4 | 1785 | |||
| XM_006243790.2 | XP_006243852.1 | 4 | 1285 | |||
| XM_006243791.2 | XP_006243853.1 | 4 | 1589 | |||
| XM_006243792.2 | XP_006243854.1 | 4 | 1167 | |||
| GenBank mRNA | BC087581.1 | - | 4 | 1007 | 7314505 | |

Ccdc51
Genomic Map