siRNA Details
Gene Symbol: Rtp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108190.1 | NP_001101660.1 | 3 | 448 | ||
XM_006243965.2 | XP_006244027.1 | 3 | 782 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rtp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108190.1 | NP_001101660.1 | 3 | 448 | ||
XM_006243965.2 | XP_006244027.1 | 3 | 782 |