siRNA Details
| Gene Symbol: CHRD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304472.1 | NP_001291401.1 | 11 | 1537 | ||
| NM_001304473.1 | NP_001291402.1 | 12 | 1563 | |||
| NM_001304474.1 | NP_001291403.1 | 11 | 1508 | |||
| NM_003741.3 | NP_003732.2 | 11 | 1537 | |||
| NR_130747.1 | 9 | 1695 | ||||
| XM_011513254.1 | XP_011511556.1 | 11 | 1390 | |||
| XM_011513256.1 | XP_011511558.1 | 9 | 1546 | |||
| XM_017007388.1 | XP_016862877.1 | 10 | 1471 | |||
| XM_017007389.1 | XP_016862878.1 | 10 | 1471 | |||
| XM_017007390.1 | XP_016862879.1 | 10 | 1471 | |||
| XM_017007391.1 | XP_016862880.1 | 10 | 1471 | |||
| XM_017007392.1 | XP_016862881.1 | 11 | 1390 | |||
| XM_017007393.1 | XP_016862882.1 | 10 | 1471 | |||
| XM_017007394.1 | XP_016862883.1 | 9 | 1259 | |||
| GenBank mRNA | AB208947.1 | - | 6 | 2088 | ||
| AF076612.1 | - | 9 | 949 | |||
| AF209928.1 | - | 11 | 1537 | |||
| AF209930.1 | - | 12 | 1563 | |||
| AF283325.1 | - | 11 | 1508 | |||
| AK074147.1 | - | 9 | 1448 | |||
| AK293456.1 | - | 11 | 1259 | |||
| AK300213.1 | - | 3 | 879 | |||
| AY358926.1 | - | 11 | 1427 | |||
| BC112345.1 | - | 11 | 1322 | 40008417 | ||

CHRD
Genomic Map