siRNA Details
Gene Symbol: CHRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304472.1 | NP_001291401.1 | 17, 18 | 2534 | ||
NM_001304473.1 | NP_001291402.1 | 18, 19 | 2560 | |||
NM_001304474.1 | NP_001291403.1 | 17, 18 | 2505 | |||
NM_003741.3 | NP_003732.2 | 17, 18 | 2534 | |||
NR_130747.1 | 15, 16 | 2692 | ||||
XM_011513254.1 | XP_011511556.1 | 16, 17 | 2123 | |||
XM_011513256.1 | XP_011511558.1 | 15, 16 | 2543 | |||
XM_017007388.1 | XP_016862877.1 | 16, 17 | 2468 | |||
XM_017007389.1 | XP_016862878.1 | 16, 17 | 2468 | |||
XM_017007390.1 | XP_016862879.1 | 15, 16 | 2204 | |||
XM_017007391.1 | XP_016862880.1 | 15, 16 | 2204 | |||
XM_017007392.1 | XP_016862881.1 | 16, 17 | 2123 | |||
XM_017007394.1 | XP_016862883.1 | 15, 16 | 2256 | |||
GenBank mRNA | AB208947.1 | - | 10, 11 | 3506 | ||
AF038198.1 | - | 4, 5 | 638 | |||
AF076612.1 | - | 14, 15 | 1826 | |||
AF209928.1 | - | 17, 18 | 2534 | |||
AF209930.1 | - | 18, 19 | 2560 | |||
AF283325.1 | - | 17, 18 | 2505 | |||
AK074147.1 | - | 15, 16 | 2445 | |||
AK293456.1 | - | 17, 18 | 2256 | |||
AK300213.1 | - | 8, 9 | 1612 | |||
AK300476.1 | - | 6, 7 | 963 | |||
AY358926.1 | - | 17, 18 | 2424 | |||
BC029879.1 | - | 4, 5 | 664 | |||
BC112345.1 | - | 17, 18 | 2319 | 40008417 |
