siRNA Details
| Gene Symbol: Xylb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033704.1 | NP_001028876.1 | 13, 14 | 1137 | ||
| XM_006244093.1 | XP_006244155.1 | 14, 15 | 1509 | |||
| XM_006244094.1 | XP_006244156.1 | 14, 15 | 1509 | |||
| XM_006244095.1 | XP_006244157.1 | 14, 15 | 1509 | |||
| XM_006244096.1 | XP_006244158.1 | 12, 13 | 1149 | |||
| GenBank mRNA | BC101852.1 | - | 13, 14 | 1137 | 7930657 | |

Xylb
Genomic Map