siRNA Details
| Gene Symbol: Trak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134565.1 | NP_001128037.1 | 2 | 244 | ||
| XM_006244100.2 | XP_006244162.1 | 2 | 251 | |||
| XM_006244101.2 | XP_006244163.1 | 2 | 3035 | |||
| XM_006244103.2 | XP_006244165.1 | 3 | 667 | |||

Trak1
Genomic Map