siRNA Details
| Gene Symbol: Zbtb47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244165.2 | XP_006244227.1 | 3 | 1721 | ||
| XM_006244166.2 | XP_006244228.1 | 3 | 1755 | |||
| XM_006244167.2 | XP_006244229.1 | 3 | 1728 | |||
| XM_006244168.2 | XP_006244230.1 | 3 | 1618 | |||
| XM_008766708.1 | XP_008764930.1 | 3 | 1563 | |||

Zbtb47
Genomic Map