siRNA Details
| Gene Symbol: Klhl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108195.1 | NP_001101665.1 | 1 | 858 | ||
| XM_006244106.2 | XP_006244168.1 | 1 | 923 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl40| Gene Symbol: Klhl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108195.1 | NP_001101665.1 | 1 | 858 | ||
| XM_006244106.2 | XP_006244168.1 | 1 | 923 | |||