siRNA Details
| Gene Symbol: Snupn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004270.1 | NP_001004270.1 | 2 | 299 | ||
| XM_006243124.1 | XP_006243186.1 | 3 | 925 | |||
| XM_006243125.1 | XP_006243187.1 | 3 | 586 | |||
| XM_006243126.1 | XP_006243188.1 | 3 | 605 | |||
| XM_006243127.2 | XP_006243189.1 | 3 | 549 | |||
| XM_006243128.2 | XP_006243190.1 | 3 | 925 | |||
| GenBank mRNA | BC079451.1 | - | 2 | 299 | 7121061 | |

Snupn
Genomic Map