siRNA Details
| Gene Symbol: Uhrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008882.1 | NP_001008882.1 | 7 | 1014 | ||
| GenBank mRNA | AY321334.1 | - | 7 | 1014 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uhrf1| Gene Symbol: Uhrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008882.1 | NP_001008882.1 | 7 | 1014 | ||
| GenBank mRNA | AY321334.1 | - | 7 | 1014 | ||