siRNA Details
| Gene Symbol: Med20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013178.2 | NP_001013196.1 | 3 | 366 | ||
| GenBank mRNA | BC083734.1 | - | 3 | 348 | 7191561 | |
| FN804985.1 | - | 3 | 328 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Med20| Gene Symbol: Med20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013178.2 | NP_001013196.1 | 3 | 366 | ||
| GenBank mRNA | BC083734.1 | - | 3 | 348 | 7191561 | |
| FN804985.1 | - | 3 | 328 | |||