siRNA Details
| Gene Symbol: Usp49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136470.1 | NP_001129942.1 | 3 | 728 | ||
| XM_006244435.2 | XP_006244497.1 | 2 | 331 | |||
| XM_008766857.1 | XP_008765079.1 | 1 | 449 | |||
| XM_008766858.1 | XP_008765080.1 | 2 | 827 | |||
| XM_008766859.1 | XP_008765081.1 | 1 | 449 | |||

Usp49
Genomic Map