siRNA Details
| Gene Symbol: Tfeb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025707.1 | NP_001020878.1 | 6 | 994 | ||
| XM_006244440.2 | XP_006244502.1 | 7 | 1923 | |||
| XM_006244442.1 | XP_006244504.1 | 7 | 913 | |||
| XM_006244446.2 | XP_006244508.1 | 7 | 2184 | |||
| GenBank mRNA | BC099102.1 | - | 6 | 994 | 7460021 | |

Tfeb
Genomic Map